Order Entry
United Kingdom
ContactUsLinkComponent
 
Methyl-3-hydroxy-2-naphthoate, Sigma-Aldrich®
Catalog # SIAL167045-5G
Supplier:  Merck
CAS Number:  
Methyl-3-hydroxy-2-naphthoate, Sigma-Aldrich®
Catalog # SIAL167045-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :202.21 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:212-936-9
CAS Number:883-99-8
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00004099