1-Dodecanol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol
CAS Number:112-53-8
:
Formula:H₃C(CH₂)₁₀CH₂OH
MDL Number:MFCD00004753
:186.34 g/mol
Merck Index:14,03405
Boiling Pt:260 °C (1013 hPa)
Melting Pt:22…24 °C
Density:0,81976 g/cm³ (20 °C)
Flash Pt:119 °C
Storage Temperature:Ambient
EINECS:203-982-0
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Solubility in EtOH:almost transparency
Specification Test Results
Frequently Bought Together