
3-Chloro-1-propanol 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-30-5
:
Formula:C₃H₇ClO
MDL Number:MFCD00002943
:94.54 g/mol
Boiling Pt:160…162 °C (760 mmHg)
Melting Pt:–20 °C
Density:1.13 g/cm³
Flash Pt:75 °C
Storage Temperature:Refrigerator
EINECS:210-992-9
UN:2849
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4500 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:0.1 % Na2CO3
Specification Test Results
Frequently Bought Together