(Triphenylphosphoranylidene)ketene, Sigma-Aldrich®
Catalog # SIAL688185-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:15596-07-3
Formula:C₂₀H₁₅OP
MDL Number:MFCD00040613