Tetrabromobisphenol A, Sigma-Aldrich®
Catalog # SIAL330396-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:543.87 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-236-9
CAS Number:79-94-7
Formula:C₁₅H₁₂Br₄O₂
MDL Number:MFCD00013962