Tetrabromobisphenol A
Catalog # EHERC17324700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:543.87 g/mol
EINECS:201-236-9
CAS Number:79-94-7
Formula:C₁₅H₁₂Br₄O₂
MDL Number:MFCD00013962