


4-Aminophenylmercury acetate, Sigma-Aldrich®
Catalog # SIALA9563-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:228-497-1
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:6283-24-5
Formula:C₈H₉HgNO₂