Glycerol monostearate
Catalog # ROTH3787.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:376.57 g/mol
EINECS:250-705-4
CAS Number:31566-31-1
Formula:C₂₁H₄₄O₅
MDL Number:MFCD00036186