1-(4-Acetylphenyl)-2-thiourea
Catalog # FLUO018156-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:194.26 g/mol
CAS Number:71680-92-7
Formula:C₉H₁₀N₂OS
MDL Number:MFCD00041188