Octyltrimethylammonium chloride, Sigma-Aldrich®
Catalog # SIAL75094-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.7838 g/mol
CAS Number:10108-86-8
Formula:C₁₁H₂₆ClN
MDL Number:MFCD00059977