2-Bromo-6-chloropyrazine
Catalog # COBBPY-7750-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.4291 g/mol
CAS Number:916791-07-6
Formula:C₄H₂BrClN₂
MDL Number:MFCD08752648