Order Entry
United Kingdom
ContactUsLinkComponent
(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate tech. 90%, Technical Grade
Toxic
Health
Irritant
(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate tech. 90%, Technical Grade
Catalog # L20227.03
CAS Number:  
Toxic
Health
Irritant
(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate tech. 90%, Technical Grade
Catalog # L20227.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
ADR:6.1,III
 :181.62 g/mol
UN:2206
 :GHS06
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:745783-72-6
Formula:C₉H₈ClNO
MDL Number:MFCD05664072