Organic reference standard, Triphenyl phosphate 500 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERXA17893000MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:115-86-6
- Description:Triphenyl phosphate 500 µg/ml in tert-butyl methyl ether
- Formula:C₁₈H₁₅O₄P
- Description:Triphenyl phosphate