Diethyl mesoxalate (diethyl ketomalonate) 95%
:609-09-6
:
:C₇H₁₀O₅
:MFCD00009121
:174.15 g/mol
:208…210 °C (760 mmHg)
:1.142 g/cm³
:>112 °C
:210-176-2
:GHS07
:Ambient
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4150 to 1.4190 (20°C, 589 nm)
- GC:≥94 %
Frequently Bought Together