Order Entry
United Kingdom
ContactUsLinkComponent
Diethyl dibutylmalonate ≥98%
Diethyl dibutylmalonate ≥98%
Catalog # L09902.14
CAS Number:  
Diethyl dibutylmalonate ≥98%
Catalog # L09902.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:150 °C (12 mmHg)
Density:0.950
 :272.39 g/mol
Storage Temperature:Ambient
EINECS:209-890-7
CAS Number:596-75-8
Formula:C₁₅H₂₈O₄
MDL Number:MFCD00026842