Indomethacin, Millipore®
Catalog # 405268-10
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
ADR:6.1,I
:357.80 g/mol
EINECS:200-186-5
UN:2811
CAS Number:53-86-1
Formula:C₁₉H₁₆ClNO₄
MDL Number:MFCD00057095