Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

L(-)-Carnitine ≥99%

L(-)-Carnitine ≥99%
Certificates
L(-)-Carnitine ≥99%
L(-)-Carnitine ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 541-15-1 Molecular Formula: C7H15NO3 Formula Weight: 161.20 Melting Point: ca 190° dec. Beilstein Reference: 4292315 Merck Reference: 14,1849 MDL No.: MFCD00038747

Synonyms:  Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
  • CAS Number:541-15-1
  • Formula:C₇H₁₅NO₃
  • MDL Number:MFCD00038747
  • Molecular Weight:161.20 g/mol
  • Merck Index:14,1849
  • Melting Pt:∼ 190 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:208-768-0
Product Family Options

Product Information