Order Entry
United Kingdom
ContactUsLinkComponent
DL-Methionine methyl ester hydrochloride
DL-Methionine methyl ester hydrochloride
Catalog # BACE4016125.0025
Supplier:  BACHEM BIOCHEMICA
CAS Number:  
DL-Methionine methyl ester hydrochloride
Catalog # BACE4016125.0025
Supplier:  BACHEM BIOCHEMICA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :199.7 g/mol
EINECS:240-285-0
CAS Number:16118-36-8
Formula:C₆H₁₃NO₂S·HCl
MDL Number:MFCD00067540