
Alcian blue 8GX, BioReagent, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:33864-99-2
:75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720
:1298.86 g/mol
EINECS:251-705-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together