Flash chromatography columns, PuriFlash®, neutral alumina
These flash chromatography columns with neutral alumina are suitable for the purification of acid sensitive compounds that would degrade if exposed to normal phase silica.
- Base material is granular neutral alumina
- pH stability ranges from 1 to 12
Frequently Bought Together