
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
: Merck

ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
: Merck
:30931-67-0
:
:C₁₈H₂₄N₆O₆S₄
:MFCD00010404
:548.68 g/mol
:M-Clarity™ quality level = MQ200
:250-396-6
:GHS07
:Refrigerator
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Sodium azide
: Merck
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Heptane (mixture of isomers), TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals