Pig L-Lactate dehydrogenase (from Muscle)
SERA27413.02
: Serva
:
- Pack type:Glass bottle for solids
- Pk:100 mg
- CAS No.:9001-60-9
- Source:Muscle
- Species:Pig
- Storage conditions:Refrigerator
- MDL No.:MFCD00131466
- Enzyme name:L-Lactate dehydrogenase
- Enzyme synonyms:LDH