Methotrexate hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: L-Amethopterin hydrate, Methylaminopterin hydrate, MTX hydrate, (4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoyl)-L-glutamic acid hydrate(1:x), Antifolan hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, (+)-Amethopterin hydrate, L(+)-Amethopterin hydrate, 4-Amino-10-methylfolic acid hydrate
CAS Number:133073-73-1
:
Formula:C₂₀H₂₂N₈O₅·xH₂O
MDL Number:MFCD00150847
Molecular Weight:454.44 g/mol (anhy)
:M-Clarity™ quality level = MQ200
EINECS:200-413-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together