E-64
Marketsource
Supplier: BACHEM BIOCHEMICA
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- BACE4027910.0005
- BACE4027910.0025
- Protein/peptide name:
- E-64
- E-64
- Protein synonyms:
- 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- CAS No.:
- 66701-25-5
- 66701-25-5
- MDL No.:
- MFCD00080261
- MFCD00080261
- Molecular formula:
- C₁₅H₂₇N₅O₅
- C₁₅H₂₇N₅O₅
- Storage conditions:
- Freezer
- Freezer
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.BACE4027910.0005Protein/peptide nameE-64Protein synonyms3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2SCAS No.66701-25-5MDL No.MFCD00080261Molecular formulaC₁₅H₂₇N₅O₅Storage conditionsFreezerMarketsource - Specifications:Marketsource
More
Specifications
Cat. No.BACE4027910.0025Protein/peptide nameE-64Protein synonyms3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2SCAS No.66701-25-5MDL No.MFCD00080261Molecular formulaC₁₅H₂₇N₅O₅Storage conditionsFreezerMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



