Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Thymidine (2'-deoxythymidine)

Thymidine (2'-deoxythymidine)

Glass bottle for solids, 1 g
Catalog # ROTH2321.3
Health
Irritant
Marketsource
Supplier:   Roth Carl
Thymidine (2'-deoxythymidine)
Catalog # ROTH2321.3
Supplier:  Roth Carl
Marketsource
CAS Number:  50-89-5
Thymidine (2'-deoxythymidine)

Glass bottle for solids, 1 g

CAS Number:  50-89-5
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  • CAS Number:50-89-5
  • Formula:C₁₀H₁₄N₂O₅
  • MDL Number:MFCD00006537
  • Molecular Weight:242.23 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-070-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.