
1-Naphthyl phosphate monosodium salt monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:81012-89-7
:
Formula:C₁₀H₈NaO₄P·1H₂O
MDL Number:MFCD00150615
:264.15 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Freezer
Frequently Bought Together