
1-Naphthyl phosphate monosodium salt monohydrate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:81012-89-7
:
Formula:C₁₀H₈NaO₄P·1H₂O
MDL Number:MFCD00150615
:264.15 g/mol
Melting Pt:189…191 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless to light yellow
- Titration with HClO4:≥98.0 %
- Impurity:≤100 ppm Free 1-Naphthol
Specification Test Results
Frequently Bought Together