Brilliant green, Certistain® for microscopy, Sigma-Aldrich®
Catalog # PFIZ1.01374
Supplier: PFIZER PROJECT
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
:
:482.65 g/mol
EINECS:211-190-1
UN:2811
:GHS07
Storage Temperature:Ambient
:1
CAS Number:633-03-4
Formula:C₂₇H₃₄N₂O₄S
MDL Number:MFCD00011880
ADR:6.1 III