Order Entry
United Kingdom
ContactUsLinkComponent
VWR®, Streptavidin (from Streptomyces avidinii)
VWR®, Streptavidin (from Streptomyces avidinii)
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Streptavidin (from Streptomyces avidinii)
Supplier:  VWR Chemicals
Certificates

Specifications

  • CAS No.:
    9013-20-1
  • Source:
    Streptomyces avidinii
  • Storage conditions:
    Freeze
  • MDL No.:
    MFCD00082035
  • Protein/peptide name:
    Streptavidin
  • Grade:
    for Biotechnology

Specifications

Related Information

Frequently Bought Together