Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Chymostatin

Marketsource
Supplier:   BACHEM BIOCHEMICA
Chymostatin
Supplier:  BACHEM BIOCHEMICA
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BACE4030514.0005
  • BACE4030514.0025
  • Protein/peptide name:
  • Chymostatin
  • Chymostatin
  • CAS No.:
  • 9076-44-2
  • 9076-44-2
  • MDL No.:
  • MFCD00071059
  • MFCD00071059
  • Storage conditions:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - £89.97
    £89.97
    1 * 5 mg
       £89.97

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BACE4030514.0005
    Protein/peptide name
    Chymostatin
    CAS No.
    9076-44-2
    MDL No.
    MFCD00071059
    Storage conditions
    Freezer
    Marketsource
  • Glass bottle for solids
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - £360.99
    £360.99
    1 * 25 mg
       £360.99

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BACE4030514.0025
    Protein/peptide name
    Chymostatin
    CAS No.
    9076-44-2
    MDL No.
    MFCD00071059
    Storage conditions
    Freezer
    Marketsource