1,3-Di-tert-butylimidazolium chloride ≥98%
Catalog # STRE07-0368-250MG
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:216.751 g/mol
:GHS07
CAS Number:157197-54-1
Formula:C₁₁H₂₁ClN₂
MDL Number:MFCD03840576