Order Entry
United Kingdom
ContactUsLinkComponent
 
Lithium methoxide 10% in methanol, Sigma-Aldrich®
Catalog # SIAL62562-100ML-F
Supplier:  Merck
CAS Number:  
Lithium methoxide 10% in methanol, Sigma-Aldrich®
Catalog # SIAL62562-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :37.98 g/mol
EINECS:212-737-7
UN:3274
ADR:3,II
CAS Number:865-34-9
Formula:CH₃LiO
MDL Number:MFCD00036357