Trihexyl(tetradecyl)phosphonium bromide, Sigma-Aldrich®
Catalog # SIAL96662-50G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:563.76 g/mol
CAS Number:654057-97-3
Formula:C₃₂H₆₈BrP
MDL Number:MFCD04117697