tert-Butylchlorodimethylsilane 1 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL372951-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:150.72 g/mol
:M-Clarity™ quality level = MQ100
EINECS:242-042-4
CAS Number:18162-48-6
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501