ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Sigma-Aldrich®
Catalog # SIALN9660-15VL
Supplier: Merck
CAS Number:
Specifications
- Pk:15 Vial
- Seq:0277
Specifications
About this item
:
:833.35 g/mol
EINECS:220-163-3
CAS Number:2646-71-1
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263