Trypsin
Catalog # PIER20233
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- CAS No.:9002-07-7
- Source:
- Species:
- Environmentally Preferable:
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin