Pig Trypsin (from Pancreas)
Catalog # A4148.0100
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Pig
- Storage conditions:–20° C
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- Enzyme activity:1:250