Order Entry
United Kingdom
ContactUsLinkComponent
Centrifuge tubes, ASTM, BLAUBRAND®, DAkks
Centrifuge tubes, ASTM, BLAUBRAND®, DAkks
Catalog # BRNDDAKKS362038
Supplier:  Brand
undefined
Centrifuge tubes, ASTM, BLAUBRAND®, DAkks
Catalog # BRNDDAKKS362038
Supplier:  Brand
Specifications
  • Closure type:
    Without cap
  • Capacity:
    100 ml
  • Description:
    Cylindrical, conical at the bottom, ASTM D 91
  • Length:
    203 mm
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3, graduated.
  • Withstands forces up to 700×g
  • Design, accuracy, etc. to meet ASTM specifications
  • With DAkks-Certifikate