

Oxytetracycline hydrochloride can be used as secondary standard
ACRO301275000
:
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
- Appearance:Yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Purity:≥80 % (as is, calculated)
:
:496.90 g/mol
:218-161-2
:GHS07
:GHS08
:Ambient
:2058-46-0
:C₂₂H₂₅ClN₂O₉
:MFCD00135815