Bradford's reagent, Biotechnology Grade for quantitative protein determination, for quantitative protein determination
Catalog # E530-1L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Abs.at peak A:0.95 - 1.6
- Abs. at peak A/Abs. at 595 nm:>1.7
- Bradford protein assay:Pass
- Characteristic pattern:Pass
- Peak A:463 nm - 467 nm
- pH (at 25 °C):0.5 - 1.5
Specification Test Results
About this item
: