
Cyclopentyl isocyanide 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68498-54-4
:
Formula:C₆H₉N
MDL Number:MFCD02664624
Boiling Pt:40 °C (11hPa)
Density:0.86 g/cm³
Storage Temperature:Freezer
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4390 to 1.4410 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
Frequently Bought Together