Order Entry
United Kingdom
ContactUsLinkComponent
Stirring paddles, HDPE, SP Bel-Art
Stirring paddles, HDPE, SP Bel-Art
Catalog # BELAH377700000
Stirring paddles, HDPE, SP Bel-Art
Catalog # BELAH377700000

Specifications

  • Description:
    Stirring paddle, with 76×152 mm paddle
  • Pk:
    1
  • Shaft Ø:
    25 mm
  • Shaft length:
    910 mm

Specifications

About this item

Stirring paddle made of HDPE, suitable for mixing liquids in large vats and tanks.