Order Entry
United Kingdom
ContactUsLinkComponent
5-Chloro-3-methylbenzo[b]thiophene
5-Chloro-3-methylbenzo[b]thiophene
Catalog # MAYBMWP00002EE
Supplier:  Thermo Scientific
CAS Number:  
5-Chloro-3-methylbenzo[b]thiophene
Catalog # MAYBMWP00002EE
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :182.67 g/mol
CAS Number:19404-18-3
Formula:C₉H₇ClS
MDL Number:MFCD00052506