tert-Butyl (2-nitroethyl)ether for synthesis, Sigma-Aldrich®
8.14278.0001
: Merck
:
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
- Solubility:(20 °C) insoluble
- Molar Mass:147.17 g/mol
- Boiling Point:30 - 35 °C (1 hPa)
- Flash Point:75 °C
- Density:1.00 g/cm3 (20 °C)
- Synonyms:2-Nitroethanol tert-butyl ether
- Hill Formula:C6H13NO3
- Chemical Formula:O2NCH2CH2OC(CH3)3
:
:M-Clarity™ quality level = MQ200
:77791-00-5
:C₆H₁₃NO₃