Order Entry
United Kingdom
ContactUsLinkComponent
 
Pesticide reference standard, Amitraz 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10230000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Amitraz 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10230000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    33089-61-1
  • Description:
    Amitraz 100 µg/ml in acetonitrile
  • Formula:
    C₁₉H₂₃N₃
  • Description:
    Amitra

Specifications

About this item