Naphthalene-[D8] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20905100CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:1146-65-2
- Description:Naphthalene-[D8] 10 µg/ml in cyclohexane
- Formula:C₁₀D₈
- Description:Naphthalene-[D8]