Ethylmagnesium bromide ∼39% (w/w) 3 mol/l in diethyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:925-90-6
:
Formula:C₂H₅BrMg
MDL Number:MFCD00000043
:133.27 g/mol
ADR:4.3,I
:M-Clarity™ quality level = MQ100
EINECS:213-127-3
UN:3399
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together