
Lithium tetrachloroaurate hydrate ≥99.99% (metals basis), Premion®
Catalog # 40429.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:3145-91-3
:
Formula:AuCl₄Li·H₂O
:345.72 g/mol (anhy)
EINECS:237-190-1
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00798533