Pesticide reference standard, Octachloronaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20425800CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2234-13-1
- Description:Octachloronaphthalene 10 µg/ml in cyclohexane
- Formula:C₁₀Cl₈
- Description:Octachloronaphthalene