Printer with battery pack, BMP53
Supplier: Brady
Certificates
About this item
The BMP53 label maker is a compact versatile printer with built-in Bluetooth® technology for wireless printing. The BMP53 portable printer has a 300 dpi print head and can print professional durable labels up to 38,10 mm wide for a wide range of applications including electrical, datacom, facility/safety and general laboratory labelling.
- Single colour printing on continuous or die cut labels
- Speed up to 25,40 mm per second
- Built-in cutter
- Easy top-loading cartridges
Delivery: Printer is supplied with a hard carrying case, rechargeable Li-on battery and AC adaptor with power cord, USB cable, drivers CD, product CD, quick start guide, MC-1500-595-WT-BK.
Frequently Bought Together